Merck
Flow Cytometry Assay for Recycling of LFA-1 in T-lymphocytes.
Bio-protocol (2018-12-05)
Katarzyna Potrzebowska, Janne Lehtonen, Malin Samuelsson, Lena Svensson
ABSTRACT
To enable cells to move forward, cell surface integrins are internalized into an endosomal compartment and subsequently intracellularly transported to be re-exposed at a new site on the cell membrane. Leukocytes are the fastest migrating cell type in the human body, which express the leukocyte-specific integrin LFA-1. Here, we describe a flow cytometry-based assay that allows the quantification of LFA-1 internalization and its re-expression on the cell surface in T lymphocytes. An advantage of using flow cytometry-based assay over biochemical methods is the low number of needed cells. This protocol can be also used to measure recycling of other receptors.
Sigma-Aldrich
Primaquine bisphosphate, 98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%