Merck
  • MondoA-Thioredoxin-Interacting Protein Axis Maintains Regulatory T-Cell Identity and Function in Colorectal Cancer Microenvironment.
MondoA-Thioredoxin-Interacting Protein Axis Maintains Regulatory T-Cell Identity and Function in Colorectal Cancer Microenvironment.
Gastroenterology (2021-04-27)
Ying Lu, Yangyang Li, Qi Liu, Na Tian, Peng Du, Fangming Zhu, Yichao Han, Xinnan Liu, Xisheng Liu, Xiao Peng, Xiaoxia Wang, Yuchen Wu, Lingfeng Tong, Yakui Li, Yemin Zhu, Lifang Wu, Ping Zhang, Ye Xu, Hanbei Chen, Bin Li, Xuemei Tong
ABSTRACT
The metabolic features and function of intratumoral regulatory T cells (Tregs) are ambiguous in colorectal cancer. Tumor-infiltrating Tregs are reprogrammed to exhibit high glucose-depleting properties and adapt to the glucose-restricted microenvironment. The glucose-responsive transcription factor MondoA is highly expressed in Tregs. However, the role of MondoA in colorectal cancer-infiltrating Tregs in response to glucose limitation remains to be elucidated. We performed studies using mice, in which MondoA was conditionally deleted in Tregs, and human colorectal cancer tissues. Seahorse and other metabolic assays were used to assess Treg metabolism. To study the role of Tregs in antitumor immunity, we used a subcutaneous MC38 colorectal cancer model and induced colitis-associated colorectal cancer in mice by azoxymethane and dextran sodium sulfate. Our analysis of single-cell RNA sequencing data of patients with colorectal cancer revealed that intratumoral Tregs featured low activity of the MondoA-thioredoxin-interacting protein (TXNIP) axis and increased glucose uptake. Although MondoA-deficient Tregs were less immune suppressive and selectively promoted T-helper (Th) cell type 1 (Th1) responses in a subcutaneous MC38 tumor model, Treg-specific MondoA knockout mice were more susceptible to azoxymethane-DSS-induced colorectal cancer. Mechanistically, suppression of the MondoA-TXNIP axis promoted glucose uptake and glycolysis, induced hyperglycolytic Th17-like Tregs, which facilitated Th17 inflammation, promoted interleukin 17A-induced of CD8+ T-cell exhaustion, and drove colorectal carcinogenesis. Blockade of interleukin 17A reduced tumor progression and minimized the susceptibility of MondoA-deficient mice to colorectal carcinogenesis. The MondoA-TXNIP axis is a critical metabolic regulator of Treg identity and function in the colorectal cancer microenvironment and a promising target for cancer therapy.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Sigma-Aldrich
2′,7′-Dichlorofluorescin Diacetate, Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) and nitric oxide (•NO) and for the determination of the degree of overall oxidative stress.