Merck
  • DJ-1 attenuates the glycation of mitochondrial complex I and complex III in the post-ischemic heart.
DJ-1 attenuates the glycation of mitochondrial complex I and complex III in the post-ischemic heart.
Scientific reports (2021-10-02)
Yvanna Pantner, Rohini Polavarapu, Lih-Shen Chin, Lian Li, Yuuki Shimizu, John W Calvert
ABSTRACT
DJ-1 is a ubiquitously expressed protein that protects cells from stress through its conversion into an active protease. Recent work found that the active form of DJ-1 was induced in the ischemic heart as an endogenous mechanism to attenuate glycative stress-the non-enzymatic glycosylation of proteins. However, specific proteins protected from glycative stress by DJ-1 are not known. Given that mitochondrial electron transport proteins have a propensity for being targets of glycative stress, we investigated if DJ-1 regulates the glycation of Complex I and Complex III after myocardial ischemia-reperfusion (I/R) injury. Initial studies found that DJ-1 localized to the mitochondria and increased its interaction with Complex I and Complex III 3 days after the onset of myocardial I/R injury. Next, we investigated the role DJ-1 plays in modulating glycative stress in the mitochondria. Analysis revealed that compared to wild-type control mice, mitochondria from DJ-1 deficient (DJ-1 KO) hearts showed increased levels of glycative stress following I/R. Additionally, Complex I and Complex III glycation were found to be at higher levels in DJ-1 KO hearts. This corresponded with reduced complex activities, as well as reduced mitochondrial oxygen consumption ant ATP synthesis in the presence of pyruvate and malate. To further determine if DJ-1 influenced the glycation of the complexes, an adenoviral approach was used to over-express the active form of DJ-1(AAV9-DJ1ΔC). Under I/R conditions, the glycation of Complex I and Complex III were attenuated in hearts treated with AAV9-DJ1ΔC. This was accompanied by improvements in complex activities, oxygen consumption, and ATP production. Together, this data suggests that cardiac DJ-1 maintains Complex I and Complex III efficiency and mitochondrial function during the recovery from I/R injury. In elucidating a specific mechanism for DJ-1's role in the post-ischemic heart, these data break new ground for potential therapeutic strategies using DJ-1 as a target.
Sigma-Aldrich
Mitochondria Isolation Kit, sufficient for 10-20 g (animal tissue), sufficient for 50 assays (2 mL), isolation of enriched mitochondrial fraction from animal tissues