Merck
  • Characterization of Human Medullary Thyroid Carcinoma Glycosphingolipids Identifies Potential Cancer Markers.
Characterization of Human Medullary Thyroid Carcinoma Glycosphingolipids Identifies Potential Cancer Markers.
International journal of molecular sciences (2021-10-14)
Karin Säljö, Anders Thornell, Chunsheng Jin, Olov Norlén, Susann Teneberg
ABSTRACT
Medullary thyroid carcinoma (MTC) accounts for only 1-2% of thyroid cancers; however, metastatic MTC is a mortal disease with no cure. In this study, glycosphingolipids were isolated from human MTCs and characterized by mass spectrometry and binding of carbohydrate recognizing ligands. The tissue distribution of selected compounds was investigated by immunohistochemistry. The amount of acid glycosphingolipids in the MTCs was higher than in the normal thyroid glands. The major acid glycosphingolipid was the GD3 ganglioside. Sulfatide and the gangliosides GM3 and GD1a were also present. The majority of the complex non-acid glycosphingolipids had type 2 (Galβ4GlcNAc) core chains, i.e., the neolactotetraosylceramide, the Lex, H type 2 and x2 pentaosylceramides, the Ley and A type 2 hexaosylceramides, and the A type 2 heptaosylceramide. There were also compounds with globo (GalαGalβ4Glc) core, i.e., globotriaosylceramide, globotetraosylceramide, the Forssman pentaosylceramide, and the Globo H hexaosylceramide. Immunohistochemistry demonstrated an extensive expression av Ley in the MTC cells and also a variable intensity and prevalence of Globo H and Lex. One individual with multiple endocrine neoplasia type 2B expressed the Forssman determinant, which is rarely found in humans. This study of human MTC glycosphingolipids identifies glycans that could serve as potential tumor-specific markers.
Sigma-Aldrich
Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Lectin from Helix pomatia, lyophilized powder, salt, free
Pricing and availability is not currently available.