Merck
  • SREBP1-Induced Glutamine Synthetase Triggers a Feedforward Loop to Upregulate SREBP1 through Sp1 O-GlcNAcylation and Augments Lipid Droplet Formation in Cancer Cells.
SREBP1-Induced Glutamine Synthetase Triggers a Feedforward Loop to Upregulate SREBP1 through Sp1 O-GlcNAcylation and Augments Lipid Droplet Formation in Cancer Cells.
International journal of molecular sciences (2021-09-29)
Jin-Wei Jhu, Jia-Bao Yan, Zou-Han Lin, Shih-Chieh Lin, I-Chen Peng
ABSTRACT
Glutamine and lipids are two important components of proliferating cancer cells. Studies have demonstrated that glutamine synthetase (GS) boosts glutamine-dependent anabolic processes for nucleotide and protein synthesis, but the role of GS in regulating lipogenesis remains unclear. This study identified that insulin and glutamine deprivation activated the lipogenic transcription factor sterol regulatory element-binding protein 1 (SREBP1) that bound to the GS promoter and increased its transcription. Notably, GS enhanced the O-linked N-acetylglucosaminylation (O-GlcNAcylation) of the specificity protein 1 (Sp1) that induced SREBP1/acetyl-CoA carboxylase 1 (ACC1) expression resulting in lipid droplet (LD) accumulation upon insulin treatment. Moreover, glutamine deprivation induced LD formation through GS-mediated O-GlcNAc-Sp1/SREBP1/ACC1 signaling and supported cell survival. These findings demonstrate that insulin and glutamine deprivation induces SREBP1 that transcriptionally activates GS, resulting in Sp1 O-GlcNAcylation. Subsequently, O-GlcNAc-Sp1 transcriptionally upregulates the expression of SREBP1, resulting in a feedforward loop that increases lipogenesis and LD formation in liver and breast cancer cells.
Sigma-Aldrich
Anti-Glutamine Synthetase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Sp1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
L-Methionine sulfoximine, suitable for cell culture
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-O-Linked N-Acetylglucosamine Antibody, clone RL2, clone RL2, from mouse