Merck
  • The epigenetic role of HTR1A antagonist in facilitaing GnRH expression for pubertal initiation control.
The epigenetic role of HTR1A antagonist in facilitaing GnRH expression for pubertal initiation control.
Molecular therapy. Nucleic acids (2021-08-31)
Shasha Zhou, Yihang Shen, Shaolian Zang, Xiaoqin Yin, Pin Li
ABSTRACT
Serotonin (5-hydroxytryptamine [5-HT]), a metabolite of tryptophan, acts on the components of the hypothalamus-hypophysis-gonad axis and induces puberty delay in mammals via 5-HT receptor 1A (HTR1A). However, the roles of HTR1A in the hypothalamus in pubertal regulation of gene expression are not fully understood. In the current study, the upregulated gonadotropin-releasing hormone (GnRH) expression in GT1-7 GnRH neuronal cells induced by the HTR1A antagonist WAY-100635 maleate was observed in vitro. Furthermore, RNA sequencing (RNA-seq) showed decreased expression of chromobox 4 (CBX4), a member of the polycomb-repressive complex 1 (PRC1), and the loss of RING2 and YY1 interaction with CBX4, suggesting the degradation of the PRC1 in GT1-7 cells treated with maleate. Chromatin immunoprecipitation sequencing (ChIP-seq) showed that the genome-wide occupancy of CBX4 and histone H2A lysine-119 ubiquitination (H2AK119ub) was compromised, especially on the promoter of GnRH. Finally, we determined that inactivation of phosphatidylinositol 3-kinase (PI3K)/Akt and mitogen-activated protein kinase (MAPK)/extracellular signal-regulated kinase (ERK) contributed to CBX4 downregulation. Taken together, we concluded that HTR1A antagonists could enhance GnRH transcription via PRC1 degradation and H2AK119ub loss driven by reduced CBX4 expression through PI3K/Akt and MAPK/ERK pathway suppression in GT1-7 cells and provided a potential epigenetic mechanism of action of HTR1A on GnRH gene expression for mammalian puberty onset.
Sigma-Aldrich
Anti-monoubiquitin H2A Antibody (Lys119), from rabbit, purified by affinity chromatography