Merck
  • Multi-transformable nanocarrier with tumor extracellular acidity-activated charge reversal, size reduction and ligand reemergence for in vitro efficient doxorubicin loading and delivery.
Multi-transformable nanocarrier with tumor extracellular acidity-activated charge reversal, size reduction and ligand reemergence for in vitro efficient doxorubicin loading and delivery.
Materials science & engineering. C, Materials for biological applications (2020-08-19)
Jumin Yang, Shuai Liu, Zhanpeng Ye, Liandong Deng, Anjie Dong, Jianhua Zhang
ABSTRACT
Various nanoparticles as drug delivery system provide significant improvements in the cancer treatment. However, their clinical success remains elusive in large part due to their inability to overcome both systemic and tumor tissue barriers. The nanosystems with nanoproperty-transformability (surface, size, stability and target) hold great promise for achieving enhanced delivery efficacy. However, currently available systems that are mainly polymer-based assemblies usually suffer from the intrinsic drawbacks of poor stability, premature leakage and low drug loading as well as limited transformability. In this study, we designed a facile strategy to build a novel multi-transformable MSNs@GO nanosystem for efficient doxorubicin (DOX) loading and delivery. This novel nanosystem was well characterized and investigated in vitro. The results indicated that the MSNs@GO can realize a very high drug loading ability due to the large pore surface area of MSNs and the demonstrated donor-acceptor (boron‑nitrogen) coordination interactions between phenylboronic acid-containing nanocarriers and electron donor-containing DOX. More importantly, the novel nanocarriers can simultaneously achieve charge reversal, size reduction and ligand reemergence by shielding/deshielding transition via acid-cleavable dynamic boronate bonds under in vitro simulated acidic microenvironment of tumor tissues, opening a new avenue for improving delivery efficiency of chemotherapeutics.
Sigma-Aldrich
3-Carboxyphenylboronic acid, ≥95%
Pricing and availability is not currently available.