Merck
  • A Novel NiFe2O4/Paper-Based Magnetoelastic Biosensor to Detect Human Serum Albumin.
A Novel NiFe2O4/Paper-Based Magnetoelastic Biosensor to Detect Human Serum Albumin.
Sensors (Basel, Switzerland) (2020-09-20)
Xing Guo, Rong Liu, Hongmei Li, Jingzhe Wang, Zhongyun Yuan, Wendong Zhang, Shengbo Sang
ABSTRACT
For the first time, a novel NiFe2O4/paper-based magnetoelastic (ME) biosensor was developed for rapid, sensitive, and portable detection of human serum albumin (HSA). Due to the uniquely magnetoelastic effect of NiFe2O4 nanoparticles and the excellent mechanical properties of the paper, the paper-based ME biosensor transforms the surface stress signal induced by the specific binding of HSA and antibody modified on the paper into the electromagnetic signal. The accumulated binding complex generates a compressive stress on the biosensor surface, resulting in a decrease in the biosensor's static magnetic permeability, which correlates to the HSA concentrations. To improve the sensitivity of the biosensor, the concentration of NiFe2O4 nanofluid and the impregnated numbers of the NiFe2O4 nanofluid-impregnated papers were optimized. The experimental results demonstrated that the biosensor exhibited a linear response to HSA concentrations ranging from 10 μg∙mL-1 to 200 μg∙mL-1, with a detection limit of 0.43 μg∙mL-1, which is significantly lower than the minimal diagnosis limit of microalbuminuria. The NiFe2O4/paper-based ME biosensor is easy to fabricate, and allows the rapid, highly-sensitive, and selective detection of HSA, providing a valuable analytical device for early monitoring and clinical diagnosis of microalbuminuria and nephropathy. This study shows the successful integration of the paper-based biosensor and the ME sensing analytical method will be a highly-sensitive, easy-to-use, disposable, and portable alternative for point-of-care monitoring.
Sigma-Aldrich
Iron nickel oxide, nanopowder, <50 nm particle size (APS), ≥98% trace metals basis
Pricing and availability is not currently available.