Merck
  • Changing Functional Signatures of Microglia along the Axis of Brain Aging.
Changing Functional Signatures of Microglia along the Axis of Brain Aging.
International journal of molecular sciences (2021-01-28)
Bianca Brawek, Maryna Skok, Olga Garaschuk
ABSTRACT
Microglia, the innate immune cells of the brain, are commonly perceived as resident macrophages of the central nervous system (CNS). This definition, however, requires further specification, as under healthy homeostatic conditions, neither morphological nor functional properties of microglia mirror those of classical macrophages. Indeed, microglia adapt exceptionally well to their microenvironment, becoming a legitimate member of the cellular brain architecture. The ramified or surveillant microglia in the young adult brain are characterized by specific morphology (small cell body and long, thin motile processes) and physiology (a unique pattern of Ca2+ signaling, responsiveness to various neurotransmitters and hormones, in addition to classic "immune" stimuli). Their numerous physiological functions far exceed and complement their immune capabilities. As the brain ages, the respective changes in the microglial microenvironment impact the functional properties of microglia, triggering further rounds of adaptation. In this review, we discuss the recent data showing how functional properties of microglia adapt to age-related changes in brain parenchyma in a sex-specific manner, with a specific focus on early changes occurring at middle age as well as some strategies counteracting the aging of microglia.
Sigma-Aldrich
GTS-21, ≥97% (HPLC)