Merck
  • Phenylacetic acid regioisomers possessing a N-difluoromethyl-1,2-dihydropyrid-2-one pharmacophore: evaluation as dual inhibitors of cyclooxygenases and 5-lipoxygenase with anti-inflammatory activity.
Phenylacetic acid regioisomers possessing a N-difluoromethyl-1,2-dihydropyrid-2-one pharmacophore: evaluation as dual inhibitors of cyclooxygenases and 5-lipoxygenase with anti-inflammatory activity.
Bioorganic & medicinal chemistry letters (2010-01-05)
Gang Yu, Morshed A Chowdhury, Khaled R A Abdellatif, Ying Dong, P N Praveen Rao, Dipankar Das, Carlos A Velázquez, Mavanur R Suresh, Edward E Knaus
ABSTRACT
A novel class of phenylacetic acid regioisomers possessing a N-difluoromethyl-1,2-dihydropyrid-2-one pharmacophore attached to its C-2, C-3 or C-4 position was designed for evaluation as anti-inflammatory (AI) agents. A number of compounds exhibited a combination of potent in vitro cyclooxygenase-2 (COX-2) and 5-lipoxygenase (5-LOX) inhibitory activities. 2-(1-Difluoromethyl-2-oxo-1,2-dihydropyridin-4-yl)phenylacetic acid (9a) exerted the most potent AI activity among this group of compounds. Molecular modeling studies showed that the N-difluoromethyl-1,2-dihydropyridin-2-one moiety present in 9a inserts into the secondary pocket present in COX-2 to confer COX-2 selectivity, and that the N-difluoromethyl-1,2-dihydropyrid-2-one group (9a) binds close to the region of the 15-LOX enzyme containing catalytic iron (His361, His366). Accordingly, the N-difluoromethyl-1,2-dihyrdopyrid-2-one moiety possesses properties that make it an attractive pharmacophore suitable for the design of dual COX-2/5-LOX inhibitory AI drugs.
Product Description
Supelco
Ibuprofen
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)