Merck
  • A Therapeutic Non-self-reactive SARS-CoV-2 Antibody Protects from Lung Pathology in a COVID-19 Hamster Model.
A Therapeutic Non-self-reactive SARS-CoV-2 Antibody Protects from Lung Pathology in a COVID-19 Hamster Model.
Cell (2020-10-16)
Jakob Kreye, S Momsen Reincke, Hans-Christian Kornau, Elisa Sánchez-Sendin, Victor Max Corman, Hejun Liu, Meng Yuan, Nicholas C Wu, Xueyong Zhu, Chang-Chun D Lee, Jakob Trimpert, Markus Höltje, Kristina Dietert, Laura Stöffler, Niels von Wardenburg, Scott van Hoof, Marie A Homeyer, Julius Hoffmann, Azza Abdelgawad, Achim D Gruber, Luca D Bertzbach, Daria Vladimirova, Lucie Y Li, Paula Charlotte Barthel, Karl Skriner, Andreas C Hocke, Stefan Hippenstiel, Martin Witzenrath, Norbert Suttorp, Florian Kurth, Christiana Franke, Matthias Endres, Dietmar Schmitz, Lara Maria Jeworowski, Anja Richter, Marie Luisa Schmidt, Tatjana Schwarz, Marcel Alexander Müller, Christian Drosten, Daniel Wendisch, Leif E Sander, Nikolaus Osterrieder, Ian A Wilson, Harald Prüss
ABSTRACT
The emergence of SARS-CoV-2 led to pandemic spread of coronavirus disease 2019 (COVID-19), manifesting with respiratory symptoms and multi-organ dysfunction. Detailed characterization of virus-neutralizing antibodies and target epitopes is needed to understand COVID-19 pathophysiology and guide immunization strategies. Among 598 human monoclonal antibodies (mAbs) from 10 COVID-19 patients, we identified 40 strongly neutralizing mAbs. The most potent mAb, CV07-209, neutralized authentic SARS-CoV-2 with an IC50 value of 3.1 ng/mL. Crystal structures of two mAbs in complex with the SARS-CoV-2 receptor-binding domain at 2.55 and 2.70 Å revealed a direct block of ACE2 attachment. Interestingly, some of the near-germline SARS-CoV-2-neutralizing mAbs reacted with mammalian self-antigens. Prophylactic and therapeutic application of CV07-209 protected hamsters from SARS-CoV-2 infection, weight loss, and lung pathology. Our results show that non-self-reactive virus-neutralizing mAbs elicited during SARS-CoV-2 infection are a promising therapeutic strategy.
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Roche
Streptavidin-POD conjugate, liquid, pkg of 1 mL
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture