Merck
  • Neonatal isolation augments social dominance by altering actin dynamics in the medial prefrontal cortex.
Neonatal isolation augments social dominance by altering actin dynamics in the medial prefrontal cortex.
Proceedings of the National Academy of Sciences of the United States of America (2016-10-30)
Hirobumi Tada, Tomoyuki Miyazaki, Kiwamu Takemoto, Kenkichi Takase, Susumu Jitsuki, Waki Nakajima, Mayu Koide, Naoko Yamamoto, Kasane Komiya, Kumiko Suyama, Akane Sano, Akiko Taguchi, Takuya Takahashi
ABSTRACT
Social separation early in life can lead to the development of impaired interpersonal relationships and profound social disorders. However, the underlying cellular and molecular mechanisms involved are largely unknown. Here, we found that isolation of neonatal rats induced glucocorticoid-dependent social dominance over nonisolated control rats in juveniles from the same litter. Furthermore, neonatal isolation inactivated the actin-depolymerizing factor (ADF)/cofilin in the juvenile medial prefrontal cortex (mPFC). Isolation-induced inactivation of ADF/cofilin increased stable actin fractions at dendritic spines in the juvenile mPFC, decreasing glutamate synaptic AMPA receptors. Expression of constitutively active ADF/cofilin in the mPFC rescued the effect of isolation on social dominance. Thus, neonatal isolation affects spines in the mPFC by reducing actin dynamics, leading to altered social behavior later in life.
Sigma-Aldrich
Anti-phospho-LIMK 1/2 (Tyr507/Thr508) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-GluR1 Antibody, clone C3T, rabbit monoclonal, culture supernatant, clone C3T, Upstate®