Merck
  • Synthesis and antioxidant properties of novel N-methyl-1,3,4-thiadiazol-2-amine and 4-methyl-2H-1,2,4-triazole-3(4H)-thione derivatives of benzimidazole class.
Synthesis and antioxidant properties of novel N-methyl-1,3,4-thiadiazol-2-amine and 4-methyl-2H-1,2,4-triazole-3(4H)-thione derivatives of benzimidazole class.
Bioorganic & medicinal chemistry (2008-03-14)
Canan Kuş, Gülgün Ayhan-Kilcigil, Süheyla Ozbey, F Betül Kaynak, Melek Kaya, Tülay Coban, Benay Can-Eke
ABSTRACT
Some novel 1-methyl-4-(2-(2-substitutedphenyl-1H-benzimidazol-1-yl)acetyl)thiosemicarbazides (16a-20a), 5-[(2-(substitutedphenyl)-1H-benzimidazol-1-yl)methyl]-N-methyl-1,3,4-thiadiazol-2-amines (17b-20b), and 5-[(2-(substitutedphenyl)-1H-benzimidazol-1-yl)methyl-4-methyl-2H-1,2,4-triazole-3(4H)-thiones (16c-20c) were synthesized and tested for antioxidant properties by using various in vitro systems. Compounds 16a-20a were found to be a good scavenger of DPPH radical (IC(50), 26 microM; IC(50), 30 microM; IC(50), 43 microM; IC(50), 55 microM; IC(50), 74 microM, respectively) when compared to BHT (IC(50), 54 microM). Noteworthy results could not be found on superoxide radical. Compound 19b, which is the most active derivative inhibited slightly lipid peroxidation (28%) at 10(-3)M concentration. Compound 17c inhibited the microsomal ethoxyresorufin O-deethylase (EROD) activity with an IC(50)=4.5 x 10(-4)M which is similarly better than the specific inhibitor caffeine IC(50)=5.2 x 10(-4)M.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg