Merck
  • Activin receptor signaling regulates cocaine-primed behavioral and morphological plasticity.
Activin receptor signaling regulates cocaine-primed behavioral and morphological plasticity.
Nature neuroscience (2015-06-02)
Amy M Gancarz, Zi-Jun Wang, Gabrielle L Schroeder, Diane Damez-Werno, Kevin M Braunscheidel, Lauren E Mueller, Monica S Humby, Aaron Caccamise, Jennifer A Martin, Karen C Dietz, Rachael L Neve, David M Dietz
ABSTRACT
Activin receptor signaling, including the transcription factor Smad3, was upregulated in the rat nucleus accumbens (NAc) shell following withdrawal from cocaine. Direct genetic and pharmacological manipulations of this pathway bidirectionally altered cocaine seeking while governing morphological plasticity in NAc neurons. Thus, Activin/Smad3 signaling is induced following withdrawal from cocaine, and such regulation may be a key molecular mechanism underlying behavioral and cellular plasticity in the brain following cocaine self-administration.
Sigma-Aldrich
PhosphoDetect Anti-Smad3 (pSer423/425) Rabbit mAb (C25A9), liquid, clone C25A9, Calbiochem®
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder