Merck
  • Genome-wide analysis of copy number alterations led to the characterisation of PDCD10 as oncogene in ovarian cancer.
Genome-wide analysis of copy number alterations led to the characterisation of PDCD10 as oncogene in ovarian cancer.
Translational oncology (2021-01-31)
Carmela De Marco, Pietro Zoppoli, Nicola Rinaldo, Sandro Morganella, Matteo Morello, Valeria Zuccalà, Maria Vincenza Carriero, Donatella Malanga, Roberta Chirillo, Paola Bruni, Carmine Malzoni, Dolores Di Vizio, Roberta Venturella, Fulvio Zullo, Antonia Rizzuto, Michele Ceccarelli, Gennaro Ciliberto, Giuseppe Viglietto
ABSTRACT
Copy Number Alterations (CNAs) represent the most common genetic alterations identified in ovarian cancer cells, being responsible for the extensive genomic instability observed in this cancer. Here we report the identification of CNAs in a cohort of Italian patients affected by ovarian cancer performed by SNP-based array. Our analysis allowed the identification of 201 significantly altered chromosomal bands (70 copy number gains; 131 copy number losses). The 3300 genes subjected to CNA identified here were compared to those present in the TCGA dataset. The analysis allowed the identification of 11 genes with increased CN and mRNA expression (PDCD10, EBAG9, NUDCD1, ENY2, CSNK2A1, TBC1D20, ZCCHC3, STARD3, C19orf12, POP4, UQCRFS1). PDCD10 was selected for further studies because of the highest frequency of CNA. PDCD10 was found, by immunostaining of three different Tissue Micro Arrays, to be over-expressed in the majority of ovarian primary cancer samples and in metastatic lesions. Moreover, significant correlations were found in specific subsets of patients, between increased PDCD10 expression and grade (p < 0.005), nodal involvement (p < 0.05) or advanced FIGO stage (p < 0.01). Finally, manipulation of PDCD10 expression by shRNA in ovarian cancer cells (OVCAR-5 and OVCA429) demonstrated a positive role for PDCD10 in the control of cell growth and motility in vitro and tumorigenicity in vivo. In conclusion, this study allowed the identification of novel genes subjected to copy number alterations in ovarian cancer. In particular, the results reported here point to a prominent role of PDCD10 as a bona fide oncogene.
Sigma-Aldrich
Anti-PDCD10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-PDCD10 (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Rho Activation Assay Kit, PP1/PP2A Toolbox for the selective in vitro dephosphorylation of proteins.