Merck
  • Small Molecules Co-targeting CKIα and the Transcriptional Kinases CDK7/9 Control AML in Preclinical Models.
Small Molecules Co-targeting CKIα and the Transcriptional Kinases CDK7/9 Control AML in Preclinical Models.
Cell (2018-08-28)
Waleed Minzel, Avanthika Venkatachalam, Avner Fink, Eric Hung, Guy Brachya, Ido Burstain, Maya Shaham, Amitai Rivlin, Itay Omer, Adar Zinger, Shlomo Elias, Eitan Winter, Paul E Erdman, Robert W Sullivan, Leah Fung, Frank Mercurio, Dansu Li, Joseph Vacca, Nathali Kaushansky, Liran Shlush, Moshe Oren, Ross Levine, Eli Pikarsky, Irit Snir-Alkalay, Yinon Ben-Neriah
ABSTRACT
CKIα ablation induces p53 activation, and CKIα degradation underlies the therapeutic effect of lenalidomide in a pre-leukemia syndrome. Here we describe the development of CKIα inhibitors, which co-target the transcriptional kinases CDK7 and CDK9, thereby augmenting CKIα-induced p53 activation and its anti-leukemic activity. Oncogene-driving super-enhancers (SEs) are highly sensitive to CDK7/9 inhibition. We identified multiple newly gained SEs in primary mouse acute myeloid leukemia (AML) cells and demonstrate that the inhibitors abolish many SEs and preferentially suppress the transcription elongation of SE-driven oncogenes. We show that blocking CKIα together with CDK7 and/or CDK9 synergistically stabilize p53, deprive leukemia cells of survival and proliferation-maintaining SE-driven oncogenes, and induce apoptosis. Leukemia progenitors are selectively eliminated by the inhibitors, explaining their therapeutic efficacy with preserved hematopoiesis and leukemia cure potential; they eradicate leukemia in MLL-AF9 and Tet2-/-;Flt3ITD AML mouse models and in several patient-derived AML xenograft models, supporting their potential efficacy in curing human leukemia.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
NVP-2, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Poly(ethylene glycol), average Mn 400
Supelco
1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®