Merck
  • Infection of Brain Organoids and 2D Cortical Neurons with SARS-CoV-2 Pseudovirus.
Infection of Brain Organoids and 2D Cortical Neurons with SARS-CoV-2 Pseudovirus.
Viruses (2020-09-12)
Sang Ah Yi, Ki Hong Nam, Jihye Yun, Dongmin Gim, Daeho Joe, Yong Ho Kim, Han-Joo Kim, Jeung-Whan Han, Jaecheol Lee
ABSTRACT
Since the global outbreak of SARS-CoV-2 (COVID-19), infections of diverse human organs along with multiple symptoms continue to be reported. However, the susceptibility of the brain to SARS-CoV-2, and the mechanisms underlying neurological infection are still elusive. Here, we utilized human embryonic stem cell-derived brain organoids and monolayer cortical neurons to investigate infection of brain with pseudotyped SARS-CoV-2 viral particles. Spike-containing SARS-CoV-2 pseudovirus infected neural layers within brain organoids. The expression of ACE2, a host cell receptor for SARS-CoV-2, was sustained during the development of brain organoids, especially in the somas of mature neurons, while remaining rare in neural stem cells. However, pseudotyped SARS-CoV-2 was observed in the axon of neurons, which lack ACE2. Neural infectivity of SARS-CoV-2 pseudovirus did not increase in proportion to viral load, but only 10% of neurons were infected. Our findings demonstrate that brain organoids provide a useful model for investigating SARS-CoV-2 entry into the human brain and elucidating the susceptibility of the brain to SARS-CoV-2.
Sigma-Aldrich
Anti-MAP-2 Antibody, from rabbit, purified by affinity chromatography