Merck
  • Immunohistochemical prediction of brain metastases in patients with advanced breast cancer: the role of Rad51.
Immunohistochemical prediction of brain metastases in patients with advanced breast cancer: the role of Rad51.
Breast (Edinburgh, Scotland) (2013-09-26)
Katarzyna Sosińska-Mielcarek, Renata Duchnowska, Piotr Winczura, Andrzej Badzio, Hanna Majewska, Joanna Lakomy, Rafał Pęksa, Beata Pieczyńska, Barbara Radecka, Sylwia Dębska, Wojciech Biernat, Jacek Jassem
ABSTRACT
There are no clinically useful biomarkers predictive of brain metastases (BM) in breast cancer. In this study, we investigated the correlation between expression of selected proteins in the primary tumor and the risk of BM in patients with metastatic breast cancer (MBC). The study included 198 MBC patients (96 with and 102 without BM). Using tissue microarrays derived from the primary tumor, we assessed by immunohistochemical expression of ER, PR, HER2, Ki-67, CK5/6, EGFR, HER3, CXCR4, Rad51, E-cadherin, and claudin 3 and 4. Ki-67 ≥14% (hazard ratio [HR] 2.76; P < 0.001), cytoplasmic expression of Rad51 (HR 1.87; P = 0.014) and ER-negativity (HR 1.72; P = 0.029) were associated with increased risk of BM in the multivariate analysis. A three-biomarker profile including ER, Ki-67 and Rad51 vs. other subtypes combined yielded an HR of 4.43 (P < 0.001). ER-negativity, cytoplasmic expression of Rad51 and high Ki-67 are associated with increased risk of BM.
Sigma-Aldrich
Anti-Cytokeratin 5 Antibody, 6, clone D5/16B4, clone D5/16B4, Chemicon®, from mouse