Merck
  • Lgr5 Marks Adult Progenitor Cells Contributing to Skeletal Muscle Regeneration and Sarcoma Formation.
Lgr5 Marks Adult Progenitor Cells Contributing to Skeletal Muscle Regeneration and Sarcoma Formation.
Cell reports (2020-12-29)
Carly Leung, Katzrin Bte Ahmad Murad, Adelyn Liang Thing Tan, Swathi Yada, Sowmya Sagiraju, Peter Karl Bode, Nick Barker
ABSTRACT
Regeneration of adult skeletal muscle is driven largely by resident satellite cells, a stem cell population increasingly considered to display a high degree of molecular heterogeneity. In this study, we find that Lgr5, a receptor for Rspo and a potent mediator of Wnt/β-catenin signaling, marks a subset of activated satellite cells that contribute to muscle regeneration. Lgr5 is found to be rapidly upregulated in purified myogenic progenitors following acute cardiotoxin-induced injury. In vivo lineage tracing using our Lgr5-2ACreERT2R26tdTomatoLSL reporter mouse model shows that Lgr5+ cells can reconstitute damaged muscle fibers following muscle injury, as well as replenish the quiescent satellite cell pool. Moreover, conditional mutation in Lgr52ACreERT2;KrasG12D;Trp53flox/flox mice drives undifferentiated pleomorphic sarcoma formation in adult mice, thereby substantiating Lgr5+ cells as a cell of origin of sarcomas. Our findings provide the groundwork for developing Rspo/Wnt-signaling-based therapeutics to potentially enhance regenerative outcomes of skeletal muscles in degenerative muscle diseases.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sigma-Aldrich
Panorama® Microbial Array Wash Solution
Sigma-Aldrich
Anti-Neurofilament H (200 kDa) Antibody, CT, serum, Chemicon®
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Slow) antibody produced in mouse, clone NOQ7.5.4D, ascites fluid