Merck
  • Identification of SARS-CoV-2 inhibitors using lung and colonic organoids.
Identification of SARS-CoV-2 inhibitors using lung and colonic organoids.
Nature (2020-10-30)
Yuling Han, Xiaohua Duan, Liuliu Yang, Benjamin E Nilsson-Payant, Pengfei Wang, Fuyu Duan, Xuming Tang, Tomer M Yaron, Tuo Zhang, Skyler Uhl, Yaron Bram, Chanel Richardson, Jiajun Zhu, Zeping Zhao, David Redmond, Sean Houghton, Duc-Huy T Nguyen, Dong Xu, Xing Wang, Jose Jessurun, Alain Borczuk, Yaoxing Huang, Jared L Johnson, Yuru Liu, Jenny Xiang, Hui Wang, Lewis C Cantley, Benjamin R tenOever, David D Ho, Fong Cheng Pan, Todd Evans, Huanhuan Joyce Chen, Robert E Schwartz, Shuibing Chen
ABSTRACT
There is an urgent need to create novel models using human disease-relevant cells to study severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) biology and to facilitate drug screening. Here, as SARS-CoV-2 primarily infects the respiratory tract, we developed a lung organoid model using human pluripotent stem cells (hPSC-LOs). The hPSC-LOs (particularly alveolar type-II-like cells) are permissive to SARS-CoV-2 infection, and showed robust induction of chemokines following SARS-CoV-2 infection, similar to what is seen in patients with COVID-19. Nearly 25% of these patients also have gastrointestinal manifestations, which are associated with worse COVID-19 outcomes1. We therefore also generated complementary hPSC-derived colonic organoids (hPSC-COs) to explore the response of colonic cells to SARS-CoV-2 infection. We found that multiple colonic cell types, especially enterocytes, express ACE2 and are permissive to SARS-CoV-2 infection. Using hPSC-LOs, we performed a high-throughput screen of drugs approved by the FDA (US Food and Drug Administration) and identified entry inhibitors of SARS-CoV-2, including imatinib, mycophenolic acid and quinacrine dihydrochloride. Treatment at physiologically relevant levels of these drugs significantly inhibited SARS-CoV-2 infection of both hPSC-LOs and hPSC-COs. Together, these data demonstrate that hPSC-LOs and hPSC-COs infected by SARS-CoV-2 can serve as disease models to study SARS-CoV-2 infection and provide a valuable resource for drug screening to identify candidate COVID-19 therapeutics.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SATB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution