Merck
  • Synthesis and structure-activity relationships of novel benzene sulfonamides with potent binding affinity for bovine carbonic anhydrase II.
Synthesis and structure-activity relationships of novel benzene sulfonamides with potent binding affinity for bovine carbonic anhydrase II.
Bioorganic & medicinal chemistry letters (2005-10-11)
Sally-Ann Poulsen, Laurent F Bornaghi, Peter C Healy
ABSTRACT
This manuscript reports the identification of a novel series of mono- and bis- benzene sulfonamides with potent binding affinity for bovine carbonic anhydrase II (bCAII). These compounds exhibited nanomolar equilibrium dissociation constants with K(i)'s ranging from 4.7 to 9.3nM. All compounds were ester derivatives of the weak affinity bCAII inhibitor, 4-carboxybenzenesulfonamide. Structure-activity relationships for this novel series of compounds are discussed.
Sigma-Aldrich
4-Sulfamoylbenzoic acid, 97%