Merck
  • Local Production of Osteoprotegerin by Osteoblasts Suppresses Bone Resorption.
Local Production of Osteoprotegerin by Osteoblasts Suppresses Bone Resorption.
Cell reports (2020-09-10)
Keisha M Cawley, Nancy Cecile Bustamante-Gomez, Anveshi G Guha, Ryan S MacLeod, Jinhu Xiong, Igor Gubrij, Yu Liu, Robin Mulkey, Michela Palmieri, Jeff D Thostenson, Joseph J Goellner, Charles A O'Brien
ABSTRACT
Osteoprotegerin (OPG) inhibits the ability of receptor activator of nuclear factor κB (NF-κB) ligand (RANKL) to stimulate the differentiation, activity, and survival of bone-resorbing osteoclasts. Genetic studies in mice show that osteocytes are an important source of RANKL, but the cellular sources of OPG are unclear. We use conditional deletion of Tnfrsf11b, which encodes OPG, from different cell populations to identify functionally relevant sources of OPG in mice. Deletion from B lymphocytes and osteocytes, two cell types commonly thought to supply OPG, has little or no impact on bone mass. By contrast, deletion of Tnfrsf11b from osteoblasts increases bone resorption and reduces bone mass to an extent similar to germline deletion, demonstrating that osteoblasts are an essential source of OPG. These results suggest that, in addition to producing new bone matrix, osteoblasts also play an active role in terminating the resorption phase of the bone remodeling cycle by suppressing RANKL activity.
Sigma-Aldrich
Human TNFRSF11A ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.