Merck
  • Sensitive detection of quinoline-derivatized sitagliptin in small volumes of human plasma by MALDI-TOF mass spectrometry.
Sensitive detection of quinoline-derivatized sitagliptin in small volumes of human plasma by MALDI-TOF mass spectrometry.
Talanta (2020-08-17)
Yi-Shan Li, Wei-Lung Tseng, Chi-Yu Lu
ABSTRACT
Dipeptidyl peptidase 4 (DPP-4) inhibitors are incretin-based medications used as oral antidiabetic agents for the treatment of type 2 diabetes. However, DPP-4 inhibitors produce side effects like acute pancreatitis, upper respiratory tract infection, nasopharyngitis, urinary tract infection, serious allergies, cardiovascular diseases, hemolysis, and retinopathy. Hence, the development of a fast and simple method to detect DPP-4 inhibitors in body fluids is important. In this study, we developed a derivatization-assisted microextraction method to enhance the detection sensitivity for trace levels of a DPP-4 inhibitor, sitagliptin, from a small volume (10 μL) of human plasma by using matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF MS). Subjecting the analyte to 100 W microwave irradiation after derivatization using a quinoline alkylating reagent (8-bromomethyl quinilone, BrMQ) shortened the reaction time to ~120 s and allowed the target analyte to be easily extracted to a small volume of the organic layer (20 μL). The analyte was then detected by MALDI-TOF MS using α-cyano-4-hydroxycinnamic acid as the matrix. The relative standard deviation and relative error were below 10% in intra- and inter-day assays. Using sitagliptin-d4 as an internal standard, the limits of quantitation and detection were found to be 0.03 μg/mL and 0.01 μg/mL, respectively. All the derivatization and extraction procedures described herein were of microliter grade. This method could effectively reduce the use of organic chemicals and solvents, thereby proving to be an eco-friendly strategy that will cause no harm to the environment.
Sigma-Aldrich
8-(Bromomethyl)quinoline, ≥96.5% (GC)
Sigma-Aldrich
2-(Bromomethyl)-6-methylpyridine, 97%