Merck
  • Functional characterization of a novel gene, Hc-dhs-28 and its role in protecting the host after Haemonchus contortus infection through regulation of diapause formation.
Functional characterization of a novel gene, Hc-dhs-28 and its role in protecting the host after Haemonchus contortus infection through regulation of diapause formation.
International journal for parasitology (2020-08-29)
Yi Yang, Xiaolu Guo, Xueqiu Chen, Jingru Zhou, Fei Wu, Yan Huang, Hengzhi Shi, Aifang Du
ABSTRACT
Haemonchus contortus could enter the diapause stage to avoid hostile conditions, however the inducing mechanism still remains poorly understood. A similar dauer strategy exists in Caenorhabditis elegans, and dauer phenomones, which are produced through a four step cycle of peroxisomal fatty acid β-oxidation, are essential in this stage. In this study, a novel gene, Hc-dhs-28, was identified and characterised. Hc-DHS-28 was the homologue of Ce-DHS-28, a key enzyme in the oxidation cycle, and the protein contained a short chain dehydrogenase domain and a peroxisomal targeting signal 1. The expression pattern of Hc-DHS-28 detected by quantitative real-time PCR and indirect immunofluorescence assay revealed that this protein was mainly expressed in the intestine and subdermal regions of larvae at diapause and in free-living stages. Enzyme activity analysis confirmed its 3-hydroxyacyl CoA dehydrogenase activity with 121, 149, 162 and 166 as key functional sites; meanwhile co-localization in human embryonic kidney 293 cells indicated that Hc-DHS-28 was targeted to the peroxisome of cytoplasm under the guide of peroxisomal targeting signal 1, which was consistent with the functional domain prediction of Hc-dhs-28. Overexpression, rescue and RNA interference experiments were carried out to explore the function of Hc-dhs-28. Our results showed that Hc-dhs-28 was very similar to Ce-dhs-28 and partially rescued its function in C. elegans. RNAi with Hc-dhs-28 in C. elegans led to decreased transcription of genes in the peroxisomal fatty acid β-oxidation cycle, considerable fat accumulation and dauer formation defects. Furthermore, immunisation with recombinant Hc-DHS-28 protein in sheep was able to maintain the body weight of the host after infection and reduce the worm burden. In conclusion, Hc-DHS-28 is most likely involved in the peroxisome fatty acid β-oxidation as the third 3-hydroxyacyl CoA dehydrogenase to regulate the production of diapause-related pheromones, and then influence the formation of diapause in H. contortus.
Sigma-Aldrich
DL-β-Hydroxybutyryl coenzyme A lithium salt, ≥90%
Pricing and availability is not currently available.