Merck
  • Pdx-1 or Pdx-1-VP16 protein transduction induces beta-cell gene expression in liver-stem WB cells.
Pdx-1 or Pdx-1-VP16 protein transduction induces beta-cell gene expression in liver-stem WB cells.
BMC research notes (2009-01-13)
Juliette Cuvelier Delisle, Lionel Martignat, Laurence Dubreil, Pierre Saï, Jean-Marie Bach, Vanessa Louzier, Steffi Bösch
ABSTRACT
Pancreatic duodenal homeobox-1 (Pdx-1) or Pdx-1-VP16 gene transfer has been shown to induce in vitro rat liver-stem WB cell conversion into pancreatic endocrine precursor cells. High glucose conditions were necessary for further differentiation into functional insulin-producing cells. Pdx-1 has the ability to permeate different cell types due to an inherent protein transduction domain (PTD). In this study, we evaluated liver-to-pancreas conversion of WB cells following Pdx-1 or Pdx-1-VP16 protein transduction. WB cells were grown in high glucose medium containing Pdx-1 or Pdx-1-VP16 recombinant proteins for two weeks. beta-like cell commitment was analysed by RT-PCR of pancreatic endocrine genes. We found that WB cells in high glucose culture spontaneously express pancreatic endocrine genes (Pdx-1, Ngn3, Nkx2.2, Kir6.2). Their further differentiation into beta-like cells expressing genes related to endocrine pancreas development (Ngn3, NeuroD, Pax4, Nkx2.2, Nkx6.1, Pdx-1) and beta-cell function (Glut-2, Kir6.2, insulin) was achieved only in the presence of Pdx-1(-VP16) protein. These results demonstrate that Pdx-1(-VP16) protein transduction is instrumental for in vitro liver-to-pancreas conversion and is an alternative to gene therapy for beta-cell engineering for diabetes cell therapy.
Sigma-Aldrich
REDTaq® DNA Polymerase, Taq for routine PCR with inert dye, 10X buffer included