Merck
  • Systemic hypoxia mimicry enhances axonal regeneration and functional recovery following peripheral nerve injury.
Systemic hypoxia mimicry enhances axonal regeneration and functional recovery following peripheral nerve injury.
Experimental neurology (2020-08-20)
Brittney D Smaila, Seth D Holland, Farshad Babaeijandaghi, Holly G Henderson, Fabio M V Rossi, Matt S Ramer
ABSTRACT
Despite the ability of peripheral nerves to regenerate after injury, failure occurs due to an inability of supporting cells to maintain growth, resulting in long-term consequences such as sensorimotor dysfunction and neuropathic pain. Here, we investigate the potential of engaging the cellular adaptive response to hypoxia, via inhibiting its negative regulators, to enhance the regenerative process. Under normoxic conditions, prolyl hydroxylase domain (PHD) proteins 1, 2, and 3 hydroxylate the key metabolic regulator hypoxia inducible factor 1α (HIF1α), marking it for subsequent proteasomal degradation. We inhibited PHD protein function systemically via either individual genetic deletion or pharmacological pan-PHD inhibition using dimethyloxalylglycine (DMOG). We show enhanced axonal regeneration after sciatic nerve crush injury in PHD1-/- mice, PHD3-/- mice, and in DMOG-treated mice, and in PHD1-/- and DMOG-treated mice a reduction in hypersensitivity to cooling after permanent sciatic ligation. Electromyographically, PHD1-/- and PHD3-/- mice showed an increased CMAP amplitude one-month post-injury, probably due to protection against denervation induced muscle atrophy, while DMOG-treated and PHD2+/- mice showed reduced latencies, indicating improved motor axon function. DMOG treatment did not affect the growth of dorsal root ganglion neurites in vitro, suggesting a lack of direct effects of DMOG on axonal regrowth. Enhanced regeneration in vivo was concurrent with an increase in macrophage density, and a shift in macrophage polarization state ratios (from M1-like toward M2-like) in DMOG-treated animals. These results indicate PHD proteins as a novel therapeutic target to improve regenerative and functional outcomes after peripheral nerve injury without manipulating molecular O2.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DMOG, ≥98% (HPLC)