Merck
  • Distance-dependent visual fluorescence immunoassay on CdTe quantum dot-impregnated paper through silver ion-exchange reaction.
Distance-dependent visual fluorescence immunoassay on CdTe quantum dot-impregnated paper through silver ion-exchange reaction.
Mikrochimica acta (2020-09-14)
Lingting Huang, Jun Wang, Qingshui Wang, Dianping Tang, Yao Lin
ABSTRACT
A paper-based visual fluorescence immunoassay is presented for the detection of matrix metalloproteinase-7 (MMP7) that is related to renal cancer. The method is based on the distance-dependent fluorescence quenching of CdTe quantum dots (QDs) on a nitrocellulose membrane by Ag+ following a sandwich-type immunoreaction on microtiter wells using silver nanoparticle (AgNP)-labeled secondary antibody- and primary antibody-coated microtiter wells. The silver nanoparticles captured in the well are dissolved with HNO3, while the quenching effect of QDs is based on silver ion-exchange reaction under 365-nm excitation light irradiation. Increasing concentration of released Ag+, thus higher concentration of the protein, leads to an increased distance of quenching on the nitrocellulose membrane. The paper-based immunoassay by combination of AgNP-assisted ion-exchange reaction with QD gives good distance-dependent responses and allows the detection of MMP7 at a concentration as low as 7.3 pg mL-1. The coefficients of variation are less than 6.9% and 12.4% for intra-assay and inter-assay, respectively. High specificity and long-term stability are achieved during the assay. Importantly, the testing of human serum samples using our strategy shows well-matched results with commercial human MMP7 ELISA kits. Graphical abstract A distance-dependent visual immunoassay is developed for the determination of serum matrix metalloproteinase-7 on CdTe quantum dot-impregnated paper with silver ion-exchange reaction.
Sigma-Aldrich
Silver, dispersion, nanoparticles, 20 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer