Merck
  • An aptamer-based colorimetric lateral flow assay for the detection of human epidermal growth factor receptor 2 (HER2).
An aptamer-based colorimetric lateral flow assay for the detection of human epidermal growth factor receptor 2 (HER2).
Analytical biochemistry (2019-10-16)
Velu Ranganathan, Sathya Srinivasan, Aryan Singh, Maria C DeRosa
ABSTRACT
An aptamer-based colorimetric lateral flow assay was developed for the detection of human epidermal growth factor receptor 2 (HER2). In this study, two approaches were examined using HER2 binding aptamers and gold nanoparticles. The first method used was a solution-based adsorption-desorption colorimetric approach wherein aptamers were adsorbed onto the gold nanoparticle surface. Upon the addition of HER2, HER2 binds specifically with its aptamer, releasing the gold nanoparticles. Addition of NaCl then induces the formation of gold nanoparticle aggregates. This leads to a color change from red to blue and a detection limit of 10 nM was achieved. The second method used an adsorption-desorption colorimetric lateral flow assay approach wherein biotin-modified aptamers were adsorbed onto the gold nanoparticle surface in the absence of HER2. In the presence of HER2, HER2 specifically binds with its aptamer leading to release of the gold nanoparticles. These solutions were applied to the lateral flow assay format and a detection limit of 20 nM was achieved. Both colorimetric and lateral flow assays are inexpensive, simple, rapid to perform and produce results visible to the naked-eye.
Product Description
Millipore
Cellulose Absorbent Sample Pads, thickness 0.83 mm, filter L × W 20 cm × 30 cm, pack of 100 sheets