Merck
Properties of Dicationic Disiloxane Ionic Liquids.
Molecules (Basel, Switzerland) (2020-07-02)
Vladimir G Krasovskiy, Gennady I Kapustin, Olga B Gorbatsevich, Lev M Glukhov, Elena A Chernikova, Anatoly A Koroteev, Leonid M Kustov
ABSTRACT
A number of dicationic ionic liquids with a disiloxane linker between imidazolium cations and bis(trifluoromethylsulfonyl)imide anion were synthesized and characterized. Melting points, viscosity, and volatility in a vacuum were measured; the thermal and hydrolytic stability of ionic liquids were also studied. The dependence of the properties on the structure of substituents in the cation of the ionic liquid was demonstrated.
Sigma-Aldrich
2-Chloroethyl methyl ether, 98%