Merck
  • Protective effects of fucoidan against 4-nitroquinolin-1-oxide provoked genetic damage in mouse bone marrow cells.
Protective effects of fucoidan against 4-nitroquinolin-1-oxide provoked genetic damage in mouse bone marrow cells.
Environmental science and pollution research international (2020-06-07)
Arumugam Ponnan, Arunkumar Kulanthaiyesu, Murugan Marudhamuthu, Kamalakkannan Palanisamy, Murugan Kadarkarai
ABSTRACT
Fucoidan is a unique bioactive and dietary polymer enriched mainly in the cell wall matrix of the brown seaweeds. This present study was intended to reveal the antigenotoxicity effect of fucoidan on 4-nitroquinolin-1-oxide (4-NQO) induced genetics damage and apoptosis in mice bone marrow cells. The 4-NQO caused genetic damages in the form of chromosome/chromatic breakage was estimated by micronuclei assay whereas apoptosis by annexin-V FITC kit and DNA damage by comet assay kit. In addition, oxidative damage in terms of plasma lipid peroxidation (LPO) and 8-OHdG was also estimated. In the experimental regime, six groups with each in five either sex of mice were used. Fucoidan constituted (50,100,200 mg/kg bwt) by orally for 5 days consequently and on 6th day, 4-NQO was administered (7.5 mg/kg bwt) by i.p. The results clearly show that negative control (H2O) and fucoidan alone constituted mice were not exhibited significant effect on LPO, genetic damages whereas positive control group (4-NQO 7.5 mg/kg bwt, i.p.) showed significant effect on genetic damage by showing increased level of LPO (6.25 vs 1.3 μM MDA), 8-OHdG (12 vs 4%), micronuclei about six-fold, 5-fold of comet, and 4-fold of apoptosis when compared with negative control, 11.6 ± 2.07, 5.00 ± 1.58, and 4.14 ± 0.65 respectively. Fucoidan pretreatment significantly protected the 4-NQO-induced genetic damage by 77% decreased level of micronuclei and 96% comet at dose of 200 mg/kg bwt over the positive control whereas LPO, 8-OHdG, and apoptosis were restored as equal to negative control. This study found as fucoidan possessing significant antigenotoxicity property by protecting 4-NQO-induced genetic damage in mice bone marrow cells as dose dependent manner suggest as valuable food supplements and medicine for mankind from environmental toxicants.
Sigma-Aldrich
May-Grünwald Stain, Suitable for the differential staining of cellular elements of blood
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
Sigma-Aldrich
Fucoidan from Undaria pinnatifida, ≥95%