Merck
  • Heterotypic cell-cell communication regulates glandular stem cell multipotency.
Heterotypic cell-cell communication regulates glandular stem cell multipotency.
Nature (2020-08-28)
Alessia Centonze, Shuheng Lin, Elisavet Tika, Alejandro Sifrim, Marco Fioramonti, Milan Malfait, Yura Song, Aline Wuidart, Jens Van Herck, Anne Dannau, Gaelle Bouvencourt, Christine Dubois, Nina Dedoncker, Amar Sahay, Viviane de Maertelaer, Christian W Siebel, Alexandra Van Keymeulen, Thierry Voet, Cédric Blanpain
ABSTRACT
Glandular epithelia, including the mammary and prostate glands, are composed of basal cells (BCs) and luminal cells (LCs)1,2. Many glandular epithelia develop from multipotent basal stem cells (BSCs) that are replaced in adult life by distinct pools of unipotent stem cells1,3-8. However, adult unipotent BSCs can reactivate multipotency under regenerative conditions and upon oncogene expression3,9-13. This suggests that an active mechanism restricts BSC multipotency under normal physiological conditions, although the nature of this mechanism is unknown. Here we show that the ablation of LCs reactivates the multipotency of BSCs from multiple epithelia both in vivo in mice and in vitro in organoids. Bulk and single-cell RNA sequencing revealed that, after LC ablation, BSCs activate a hybrid basal and luminal cell differentiation program before giving rise to LCs-reminiscent of the genetic program that regulates multipotency during embryonic development7. By predicting ligand-receptor pairs from single-cell data14, we find that TNF-which is secreted by LCs-restricts BC multipotency under normal physiological conditions. By contrast, the Notch, Wnt and EGFR pathways were activated in BSCs and their progeny after LC ablation; blocking these pathways, or stimulating the TNF pathway, inhibited regeneration-induced BC multipotency. Our study demonstrates that heterotypic communication between LCs and BCs is essential to maintain lineage fidelity in glandular epithelial stem cells.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
CCR2 Antagonist, The CCR2 Antagonist, also referenced under CAS 445479-97-0, controls the biological activity of CCR2.
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone MB1.2, clone MB1.2, Chemicon®, from rat