MilliporeSigma
  • EPHA5 mutation impairs natural killer cell-mediated cytotoxicity against non-small lung cancer cells and promotes cancer cell migration and invasion.
EPHA5 mutation impairs natural killer cell-mediated cytotoxicity against non-small lung cancer cells and promotes cancer cell migration and invasion.
Molecular and cellular probes (2020-04-03)
Jingwen Zhang, Zhihao Zhang, Weiwei Song, Jumin Liu
ABSTRACT
This study aims to evaluate the role of the EPHA5 mutation in the migration and invasion of non-small cell lung cancer (NSCLC) cells and in modulating the killing effect of natural killer (NK) cells to NSCLC cells. EPHA5-wt (wild type) and EPHA5-mut (mutation) plasmids were constructed. EPHA5 was silenced using si-EPHA5. NSCLC cell migration and invasion were determined using Transwell assays. NK cell proliferation and apoptosis were determined using CCK-8 assay and flow cytometry, respectively. The killing effect of NK cells to NSCLC cells was also examined. EPHA5 mutation significantly promoted migration and invasion in NSCLC cells. Furthermore, EPHA5 mutation notably impaired the cytotoxicity of NK cells against NSCLC cells. In contrast, EPHA5-wt overexpression and EPHA5 silencing exerted the opposite effect. EPHA5 mutation impairs the NK cell-mediated cytotoxicity against NSCLC cells and promotes migration and invasion in NSCLC cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human EPHA5