Merck
Development of a multi-material stereolithography 3D printing device.
Micromachines (2020-05-28)
Bilal Khatri, Marco Frey, Ahmed Raouf-Fahmy, Marc-Vincent Scharla, Thomas Hanemann
ABSTRACT
Additive manufacturing, or nowadays more popularly entitled as 3D printing, enables a fast realization of polymer, metal, ceramic or composite devices, which often cannot be fabricated with conventional methods. One critical issue for a continuation of this success story is the generation of multi-material devices. Whilst in fused filament fabrication or 3D InkJet printing, commercial solutions have been realized, in stereolithography only very few attempts have been seen. In this work, a comprehensive approach, covering the construction, material development, software control and multi-material printing is presented for the fabrication of structural details in the micrometer range. The work concludes with a critical evaluation and possible improvements.
Sigma-Aldrich
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide, 97%
Sigma-Aldrich
Bisphenol A glycerolate dimethacrylate, glycerol/phenol 1