Merck
  • Suppression of CaMKIIβ Inhibits ANO1-Mediated Glioblastoma Progression.
Suppression of CaMKIIβ Inhibits ANO1-Mediated Glioblastoma Progression.
Cells (2020-05-03)
Kyoung Mi Sim, Young-Sun Lee, Hee Jin Kim, Chang-Hoon Cho, Gwan-Su Yi, Myung-Jin Park, Eun Mi Hwang, Jae-Yong Park
ABSTRACT
ANO1, a Ca2+-activated chloride channel, is highly expressed in glioblastoma cells and its surface expression is involved in their migration and invasion. However, the regulation of ANO1 surface expression in glioblastoma cells is largely unknown. In this study, we found that Ca2+/Calmodulin-dependent protein kinase II (CaMKII) β specifically enhances the surface expression and channel activity of ANO1 in U251 glioblastoma cells. When KN-93, a CaMKII inhibitor, was used to treat U251 cells, the surface expression and channel activity of ANO1 were significantly reduced. Only CaMKIIβ, among the four CaMKII isoforms, increased the surface expression and channel activity of ANO1 in a heterologous expression system. Additionally, gene silencing of CaMKIIβ suppressed the surface expression and channel activity of ANO1 in U251 cells. Moreover, gene silencing of CaMKIIβ or ANO1 prominently reduced the migration and invasion of U251 and U87 MG glioblastoma cells. We thus conclude that CaMKIIβ plays a specific role in the surface expression of ANO1 and in the ANO1-mediated tumorigenic properties of glioblastoma cells, such as migration and invasion.
Sigma-Aldrich
MISSION® esiRNA, targeting human FIGN
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Sigma-Aldrich
KN-93, A cell-permeable, reversible and competitive inhibitor of rat brain CaM kinase II (Ki = 370 nM).