MilliporeSigma
  • Andrographolide modulates HNF4α activity imparting on hepatic metabolism.
Andrographolide modulates HNF4α activity imparting on hepatic metabolism.
Molecular and cellular endocrinology (2020-05-19)
Minyi Zhang, Meng Yang, Na Wang, Qingli Liu, Binxu Wang, Tongling Huang, Yan Tong, Yanlin Ming, Chi-Wai Wong, Jinsong Liu, Dongsheng Yao, Min Guan
ABSTRACT
Hepatic nuclear factor 4 alpha (HNF4α) drives the expression of apolipoprotein B (ApoB), microsomal triglyceride transfer protein (MTP) and phospholipase A2 G12B (PLA2G12B), governing hepatic very-low-density lipoprotein (VLDL) production and secretion. Andrographolide (AP) is a major constituent isolated from Andrographis paniculata. We found that AP can disrupt the interaction between HNF4α and its coactivator peroxisome proliferator-activated receptor gamma coactivator 1-alpha (PGC1α). Virtual docking and mutational analysis indicated that arginine 235 of HNF4α is essential for binding to AP. As a consequence of antagonizing the activity of HNF4α, AP suppresses the expression of ApoB, MTP and PLA2G12B and reduces the rate of hepatic VLDL secretion in vivo. AP additionally reduced gluconeogenesis via down-regulating the expression of HNF4α target genes phosphoenolpyruvate carboxykinase (Pepck) and glucose-6-phosphatase (G6pc). Collectively, our results suggest that AP affects liver function via modulating the transcriptional activity of HNF4α.
Sigma-Aldrich
T0901317, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mayer′s Hematoxylin Solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tyloxapol, nonionic surfactant
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Andrographolide, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Eosin Y Solution, Aqueous
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%