Merck
  • Oncogenic hijacking of a developmental transcription factor evokes vulnerability toward oxidative stress in Ewing sarcoma.
Oncogenic hijacking of a developmental transcription factor evokes vulnerability toward oxidative stress in Ewing sarcoma.
Nature communications (2020-05-18)
Aruna Marchetto, Shunya Ohmura, Martin F Orth, Maximilian M L Knott, Maria V Colombo, Chiara Arrigoni, Victor Bardinet, David Saucier, Fabienne S Wehweck, Jing Li, Stefanie Stein, Julia S Gerke, Michaela C Baldauf, Julian Musa, Marlene Dallmayer, Laura Romero-Pérez, Tilman L B Hölting, James F Amatruda, Andrea Cossarizza, Anton G Henssen, Thomas Kirchner, Matteo Moretti, Florencia Cidre-Aranaz, Giuseppina Sannino, Thomas G P Grünewald
ABSTRACT
Ewing sarcoma (EwS) is an aggressive childhood cancer likely originating from mesenchymal stem cells or osteo-chondrogenic progenitors. It is characterized by fusion oncoproteins involving EWSR1 and variable members of the ETS-family of transcription factors (in 85% FLI1). EWSR1-FLI1 can induce target genes by using GGAA-microsatellites as enhancers.Here, we show that EWSR1-FLI1 hijacks the developmental transcription factor SOX6 - a physiological driver of proliferation of osteo-chondrogenic progenitors - by binding to an intronic GGAA-microsatellite, which promotes EwS growth in vitro and in vivo. Through integration of transcriptome-profiling, published drug-screening data, and functional in vitro and in vivo experiments including 3D and PDX models, we discover that constitutively high SOX6 expression promotes elevated levels of oxidative stress that create a therapeutic vulnerability toward the oxidative stress-inducing drug Elesclomol.Collectively, our results exemplify how aberrant activation of a developmental transcription factor by a dominant oncogene can promote malignancy, but provide opportunities for targeted therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human SOX6
Sigma-Aldrich
MISSION® esiRNA, targeting human TXNIP