Merck
  • Correlation of Physicochemical and Antimicrobial Properties of Liposomes Loaded with (+)-Usnic Acid.
Correlation of Physicochemical and Antimicrobial Properties of Liposomes Loaded with (+)-Usnic Acid.
ChemPlusChem (2020-05-19)
Sara Battista, Pierangelo Bellio, Giuseppe Celenza, Luciano Galantini, Irene Franceschini, Giovanna Mancini, Luisa Giansanti
ABSTRACT
(+)-Usnic acid (UA) is a natural substance that displays pharmacological activity, but it is barely soluble in water, so it was included in liposomes in order to study its properties. First, the effects of phospholipid structure and loading methodology on UA entrapment efficacy were evaluated. Then, the physicochemical and biological properties (UA delivery efficacy to Staphylococcus aureus bacterial cells) of different liposome formulations containing structurally related amphiphiles derived from L-prolinol were fully investigated. Entrapment efficiency of UA with passive loading by incubation was 80-100 molar percentage, which is related to lipophilicity of the drug and to the packing and fluidity of the bilayer. Some of the investigated formulations show the potential of UA in delivery systems (minimum inhibitory concentration of liposomal UA: 8 μg/mL) and even subtle variations of the molecular structure of lipids can significantly affect the liposomes' physicochemical properties and efficiency of drug release.
Product Description
Millipore
MRSA Selective Supplement, suitable for microbiology
Sigma-Aldrich
Dialysis tubing cellulose membrane, avg. flat width 43 mm (1.7 in.)