Merck
Pd-catalyzed intermolecular C-H amination with alkylamines.
Journal of the American Chemical Society (2011-04-28)
Eun Jeong Yoo, Sandy Ma, Tian-Sheng Mei, Kelvin S L Chan, Jin-Quan Yu
ABSTRACT
C-H amination of N-aryl benzamides with O-benzoyl hydroxylamines has been achieved with either Pd(II) or Pd(0) catalysts. Furthermore, we demonstrate that secondary amines can be directly used with benzoyl peroxide in a one-pot procedure that proceeds via the in situ generation of the appropriate O-benzoyl hydroxylamines. This catalytic reaction provides a new disconnection for the convergent synthesis of tertiary and secondary arylalkyl amines starting from benzoic acids.
Sigma-Aldrich
Yu-Wasa Auxiliary, 97%