Merck
  • Ubiquitination of interleukin-1α is associated with increased pro-inflammatory polarization of murine macrophages deficient in the E3 ligase ITCH.
Ubiquitination of interleukin-1α is associated with increased pro-inflammatory polarization of murine macrophages deficient in the E3 ligase ITCH.
The Journal of biological chemistry (2020-06-27)
Xi Lin, Hengwei Zhang, Brendan F Boyce, Lianping Xing
ABSTRACT
Macrophages play critical roles in homeostasis and inflammation. Macrophage polarization to either a pro-inflammatory or anti-inflammatory status is controlled by activating inflammatory signaling pathways. Ubiquitination is a posttranslational modification that regulates these inflammatory signaling pathways. However, the influence of protein ubiquitination on macrophage polarization has not been well studied. We hypothesized that the ubiquitination status of key proteins in inflammatory pathways contributes to macrophage polarization, which is regulated by itchy E3 ubiquitin ligase (ITCH), a negative regulator of inflammation. Using ubiquitin proteomics, we found that ubiquitination profiles are different among polarized murine macrophage subsets. Interestingly, interleukin-1α (IL-1α), an important pro-inflammatory mediator, was specifically ubiquitinated in lipopolysaccharide-induced pro-inflammatory macrophages, which was enhanced in ITCH-deficient macrophages. The ITCH-deficient macrophages had increased levels of the mature form of IL-1α and exhibited pro-inflammatory polarization, and reduced deubiquitination of IL-1α protein. Finally, IL-1α neutralization attenuated pro-inflammatory polarization of the ITCH-deficient macrophages. In conclusion, ubiquitination of IL-1α is associated with increased pro-inflammatory polarization of macrophages deficient in the E3 ligase ITCH.
Product Description
Millipore
EZview Red Protein A Affinity Gel
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Millipore
EZview Red Protein G Affinity Gel