Merck
  • Nintedanib (BIBF-1120) inhibits hepatocellular carcinoma growth independent of angiokinase activity.
Nintedanib (BIBF-1120) inhibits hepatocellular carcinoma growth independent of angiokinase activity.
Journal of hepatology (2014-03-25)
Wei-Tien Tai, Chung-Wai Shiau, Yong-Shi Li, Chun-Wei Chang, Jui-Wen Huang, Ting-Ting Hsueh, Hui-Chuan Yu, Kuen-Feng Chen
ABSTRACT
Nintedanib, a triple angiokinase inhibitor, is currently being evaluated against advanced HCC in phase I/II clinical trials. Here, we report the underlying molecular mechanism by which nintedanib (BIBF-1120) induces an anti-HCC effect. To further elucidate whether the effect of nintedanib on SHP-1 is dependent on its angiokinase inhibition activity, we developed a novel kinase-independent derivative of nintedanib, ΔN. HCC cell lines were treated with nintedanib or its derivative (ΔN) and apoptosis, signal transduction, and phosphatase activity were analyzed. Purified SHP-1 proteins or HCC cells expressing deletion N-SH2 domain or D61A point mutants were used to investigate the potential effect of nintedanib on SHP-1. In vivo efficacy was determined in nude mice with HCC subcutaneous xenografts (n⩾8 mice). Nintedanib induced anti-proliferation in HCC cell lines by targeting STAT3. Ectopic STAT3 abolished nintedanib-mediated apoptosis in HCC cells. Nintedanib further activated SHP-1 in purified SHP-1 proteins suggesting that nintedanib directly affects SHP-1 for STAT3 inhibition. HCC cells or recombinant SHP-1 proteins expressing deletion of N-SH2 domain or D61A mutants restored the activity of nintedanib suggesting that the auto-inhibition structure of SHP-1 was relieved by nintedanib. Although ΔN only retained the backbone of nintedanib without kinase activity, ΔN still induced substantial anti-HCC activity in vitro and in vivo by targeting STAT3. Nintedanib induced significant anti-HCC activity independent of angiokinase inhibition activity in a preclinical HCC model by relieving autoinhibition of SHP-1. Our findings provide new mechanistic insight into the inhibition of HCC growth by nintedanib.
Sigma-Aldrich
Nintedanib ethanesulphonate, ≥98% (HPLC)
Sigma-Aldrich
Nintedanib, ≥98% (HPLC)