Merck
  • A multi-residue method for the determination of 124 pesticides in rice by modified QuEChERS extraction and gas chromatography-tandem mass spectrometry.
A multi-residue method for the determination of 124 pesticides in rice by modified QuEChERS extraction and gas chromatography-tandem mass spectrometry.
Food chemistry (2013-02-16)
Xue Hou, Mei Han, XiaoHang Dai, XiaoFeng Yang, Shengguo Yi
ABSTRACT
A modified QuEChERS method for simultaneous determination of 124 pesticide residues in rice was established and validated. The rice samples were initially extracted with acetonitrile (MeCN), and the targeted pesticides were purified following the dispersive solid phase extraction (d-SPE) cleanup method and detected by gas chromatography-tandem mass spectrometry (GC-MS/MS). The efficiency of citrate-buffered and unbuffered method in liquid extraction procedure was compared; the amount of primary secondary amine (PSA) was optimised. In the method validation, correlation coefficients (r(2)) were higher than 0.990 with the linear ranging from 10 to 200 μg kg(-1). At the fortification levels of 20-200 μg kg(-1), average recoveries ranged from 70% to 122.7% with the RSD <20%.
Product Description
Supelco
Chlorobenzilate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
Flucythrinate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.