Merck
  • Discovery of MD-224 as a First-in-Class, Highly Potent, and Efficacious Proteolysis Targeting Chimera Murine Double Minute 2 Degrader Capable of Achieving Complete and Durable Tumor Regression.
Discovery of MD-224 as a First-in-Class, Highly Potent, and Efficacious Proteolysis Targeting Chimera Murine Double Minute 2 Degrader Capable of Achieving Complete and Durable Tumor Regression.
Journal of medicinal chemistry (2018-12-12)
Yangbing Li, Jiuling Yang, Angelo Aguilar, Donna McEachern, Sally Przybranowski, Liu Liu, Chao-Yie Yang, Mi Wang, Xin Han, Shaomeng Wang
ABSTRACT
Human murine double minute 2 (MDM2) protein is a primary endogenous cellular inhibitor of the tumor suppressor p53 and has been pursued as an attractive cancer therapeutic target. Several potent, nonpeptide, small-molecule inhibitors of MDM2 are currently in clinical development. In this paper, we report our design, synthesis, and evaluation of small-molecule MDM2 degraders based on the proteolysis targeting chimera (PROTAC) concept. The most promising compound (MD-224) effectively induces rapid degradation of MDM2 at concentrations <1 nM in human leukemia cells. It achieves an IC50 value of 1.5 nM in inhibition of growth of RS4;11 cells and also low nanomolar IC50 values in a panel of leukemia cell lines. MD-224 achieves complete and durable tumor regression in vivo in the RS4;11 xenograft tumor model in mice at well-tolerated dose schedules. MD-224 is thus a highly potent and efficacious MDM2 degrader and warrants extensive evaluations as a new class of anticancer agent.
Sigma-Aldrich
Pomalidomide-C3-NH2 hydrochloride, ≥95%
Sigma-Aldrich
Pomalidomide-C6-NH2 hydrochloride, ≥95%
Sigma-Aldrich
(S,R,S)-AHPC-Me, 95%