Merck
  • Veterinary utility of dried blood spots for detailed analysis of chlorinated pesticides and polychlorinated biphenyls by gas chromatography tandem mass spectrometry.
Veterinary utility of dried blood spots for detailed analysis of chlorinated pesticides and polychlorinated biphenyls by gas chromatography tandem mass spectrometry.
Toxicology mechanisms and methods (2020-01-30)
Andreas F Lehner, Lauren Stensen, Alan Zimmerman, Adam Bush, John Buchweitz
ABSTRACT
Persistent organic pollutants (POPs) are organic compounds of anthropogenic origin that resist atmospheric and microbial degradation and thus persist in the environment and in food chains for exceptionally long periods of time. Veterinarians and wildlife researchers need simple methodologies for monitoring and measuring such compounds including two large and diverse categories, organochlorine pesticides (OCs) and polychlorinated biphenyls (PCBs), compounds that have been largely banned from production and use except for specific exceptions. We present development of methodologies for detection and quantitation of 22 OCs and 10 PCB congeners by tandem quadrupole gas chromatography-mass spectrometric analysis of Dried Blood Spots (DBS). Development was enabled by (1) optimization of suspension and extraction methodologies for DBS; (2) strategic streamlining and condensation of Multiple Reaction Monitoring (MRM) settings on GC/MS/MS; and (3) improvement of GC settings to accommodate all 32 compounds in a single chromatographic run per sample. The method was validated for parameters of linearity, limits of detection and quantitation, recovery and precision, and results from blood were shown to correlate well with those from DBS despite both being only 50 uL in volume. The method was applied successfully to blood samples from nine avian specimens submitted to the MSU Veterinary Diagnostic Lab, and all were shown to bear the burden of varying levels of OCs and/or PCB compounds.
Sigma-Aldrich
2-Methylheptane, 98%