Merck
  • Dietary exposure assessment of sum deoxynivalenol forms, sum T-2/HT-2 toxins and zearalenone from cereal-based foods and beer.
Dietary exposure assessment of sum deoxynivalenol forms, sum T-2/HT-2 toxins and zearalenone from cereal-based foods and beer.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-03-22)
Vladimir Ostry, Marcela Dofkova, Jitka Blahova, Frantisek Malir, Radek Kavrik, Irena Rehurkova, Jiri Ruprich
ABSTRACT
A dietary exposure assessment to sum of deoxynivalenol (DON) forms, sum of T-2/HT-2 toxins (T2/HT2) and zearalenone (ZEA) was conducted for Czech children 4-6 years and Czech men and women 18-59 years. Retail foods (25 different commodities, n = 336) were assessed by LC-MS/MS methods. The 95th percentile chronic exposure to sum of DON forms was determined in children from 648 to 1030 ng/kg bw/day (LB/lower bound/and UB/upper bound/), in men from 362 to 923 ng/kg bw/day and in women from 272 to 490 ng/kg bw/day. The 95th percentile chronic exposure to sum T2/HT2 was determined in children from 6.5 to 31 ng/kg bw/day, in men from 1.9 to 11.2 ng/kg bw/day and in women from 2.5 to 11.5 ng/kg bw/day. The 95th percentile chronic exposure to ZEA was determined in children from 11.9 to 24.9 ng/kg bw/day, in men from 5.9 to 27.5 ng/kg bw/day and in women from 4.8 to 12.6 ng/kg bw/day. The risk linked with the mean and the 95th percentile chronic exposure (LB scenario) to the sum of DON forms, sum of T2/HT2 and ZEA is considered to be out of health concern for the selected population groups.
Sigma-Aldrich
6-Diazo-5-oxo-L-norleucine, crystalline
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Supelco
Deoxynivalenol-13C15 solution, ~25 μg/mL in acetonitrile, analytical standard