Merck
  • STAT3 inhibition induced temozolomide-resistant glioblastoma apoptosis via triggering mitochondrial STAT3 translocation and respiratory chain dysfunction.
STAT3 inhibition induced temozolomide-resistant glioblastoma apoptosis via triggering mitochondrial STAT3 translocation and respiratory chain dysfunction.
Cellular signalling (2020-03-14)
Ping Cui, Fen Wei, Jingjing Hou, Ying Su, Jijun Wang, Sicen Wang
ABSTRACT
Recent evidence has demonstrated that the signal transducer and activator of transcription 3 (STAT3) gene are abnormally active in glioblastoma multiforme (GBM), and this change is crucial for the tumor survival and chemotherapy-resistant. Certain preclinical pharmacology studies have focused on STAT3 phosphorylation and homodimerization, and have developed a class of salicylic acid-based inhibitors, which blocks the nuclear translocation-dependent canonical STAT3 signaling. In the present study, we demonstrated that the salicylic acid-based compound SH-4-54 was quite toxic to temozolomide (TMZ)-resistant GBM cells and could trigger apoptosis in these cells via enhancing mitochondrial translocation-dependent non-canonical STAT3 pathway. We demonstrated that incubation of TMZ-resistant GBM cells with SH-4-54 led to mitochondrial STAT3 (mitoSTAT3) activation and respiratory dysfunction reflected by disrupted (or suppressed) activities of oxidative phosphorylation complexes and oxygen consumption rate. Mechanistically, we proved that SH-4-54 could increase mitoSTAT3 transmembrane import via GRIM-19 and reinforce the association between mitoSTAT3 and mitochondrial transcription factor A (TFAM), indicating that SH-4-54 could facilitate the binding of mitoSTAT3 to mitochondria DNA (mtDNA) and negatively regulate mitochondrial-encoded genes, thus leading to the abnormal oxidation respiratory. Lastly, using GRIM-19 knockout cell line and subcutaneous xenotransplanted tumor model, we elaborately showed the enrichment of SH-4-54 in mitochondria by LC-MS/MS analysis. In conclusion, our data demonstrate thatthe salicylic acid-based compound SH-4-54 is quite effective in killing TMZ-resistant GBM cells and this cytotoxicity is attributed to mitoSTAT3 activation.
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
MISSION® esiRNA, targeting human NDUFA13