Merck
  • IRF5 genetic risk variants drive myeloid-specific IRF5 hyperactivation and presymptomatic SLE.
IRF5 genetic risk variants drive myeloid-specific IRF5 hyperactivation and presymptomatic SLE.
JCI insight (2019-12-27)
Dan Li, Bharati Matta, Su Song, Victoria Nelson, Kirsten Diggins, Kim R Simpfendorfer, Peter K Gregersen, Peter Linsley, Betsy J Barnes
ABSTRACT
Genetic variants within or near the interferon regulatory factor 5 (IRF5) locus associate with systemic lupus erythematosus (SLE) across ancestral groups. The major IRF5-SLE risk haplotype is common across populations, yet immune functions for the risk haplotype are undefined. We characterized the global immune phenotype of healthy donors homozygous for the major risk and nonrisk haplotypes and identified cell lineage-specific alterations that mimic presymptomatic SLE. Contrary to previous studies in B lymphoblastoid cell lines and SLE immune cells, IRF5 genetic variants had little effect on IRF5 protein levels in healthy donors. Instead, we detected basal IRF5 hyperactivation in the myeloid compartment of risk donors that drives the SLE immune phenotype. Risk donors were anti-nuclear antibody positive with anti-Ro and -MPO specificity, had increased circulating plasma cells and plasmacytoid dendritic cells, and had enhanced spontaneous NETosis. The IRF5-SLE immune phenotype was conserved over time and probed mechanistically by ex vivo coculture, indicating that risk neutrophils are drivers of the global immune phenotype. RNA-Seq of risk neutrophils revealed increased IRF5 transcript expression, IFN pathway enrichment, and decreased expression of ROS pathway genes. Altogether, the data support that individuals carrying the IRF5-SLE risk haplotype are more susceptible to environmental/stochastic influences that trigger chronic immune activation, predisposing to the development of clinical SLE.
Product Description
Millipore
MILLIPLEX® Human Autoimmune Autoantibody Panel, Milliplex®
Pricing and availability is not currently available.
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Pricing and availability is not currently available.