Merck
  • Synthesis of C12-Keto Saxitoxin Derivatives with Unusual Inhibitory Activity Against Voltage-Gated Sodium Channels.
Synthesis of C12-Keto Saxitoxin Derivatives with Unusual Inhibitory Activity Against Voltage-Gated Sodium Channels.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-11-27)
Kanna Adachi, Tomoshi Yamada, Hayate Ishizuka, Mana Oki, Shunsuke Tsunogae, Noriko Shimada, Osamu Chiba, Tatsuya Orihara, Masafumi Hidaka, Takatsugu Hirokawa, Minami Odagi, Keiichi Konoki, Mari Yotsu-Yamashita, Kazuo Nagasawa
ABSTRACT
A novel series of C12-keto-type saxitoxin (STX) derivatives bearing an unusual nonhydrated form of the ketone at C12 has been synthesized, and their NaV -inhibitory activity has been evaluated in a cell-based assay as well as whole-cell patch-clamp recording. Among these compounds, 11-benzylidene STX (3 a) showed potent inhibitory activity against neuroblastoma Neuro 2A in both cell-based and electrophysiological analyses, with EC50 and IC50 values of 8.5 and 30.7 nm, respectively. Interestingly, the compound showed potent inhibitory activity against tetrodotoxin-resistant subtype of NaV 1.5, with an IC50 value of 94.1 nm. Derivatives 3 a-d and 3 f showed low recovery rates from NaV 1.2 subtype (ca 45-79 %) compared to natural dcSTX (2), strongly suggesting an irreversible mode of interaction. We propose an interaction model for the C12-keto derivatives with NaV in which the enone moiety in the STX derivatives 3 works as Michael acceptor for the carboxylate of Asp1717 .
Product Description
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Veratridine, ≥90% (HPLC), powder
Sigma-Aldrich
S(−)-Verapamil hydrochloride hydrate, ≥98% (HPLC), powder