Merck
  • Liquid-phase peptide synthesis on polyethylene glycol (PEG) supports using strategies based on the 9-fluorenylmethoxycarbonyl amino protecting group: application of PEGylated peptides in biochemical assays.
Liquid-phase peptide synthesis on polyethylene glycol (PEG) supports using strategies based on the 9-fluorenylmethoxycarbonyl amino protecting group: application of PEGylated peptides in biochemical assays.
Journal of peptide science : an official publication of the European Peptide Society (2002-10-10)
Peter M Fischer, Daniella I Zheleva
ABSTRACT
Stepwise synthetic assembly of polypeptide chains reversibly linked to polyethylene glycol represents a hybrid between traditional solution and solid-phase chemistries and combines the inherent advantages of both approaches. The technical simplicity and scalability of the liquid-phase peptide synthesis method renders it particularly attractive for multiple parallel syntheses, combinatorial approaches and the large-scale preparation of peptides. The versatile protection strategy based on the N alpha-fluorenylmethoxycarbonyl group commonly used in solid-phase peptide synthesis was adapted to the liquid-phase approach. Fluoride ions were used rather than the conventional organic base piperidine for the repetitive amino-deprotection step. Using a range of acid- and base-labile linkers between the polymer and the peptide, it was shown that free and fully side-chain protected peptides can be obtained using our version of the liquid-phase peptide synthesis method. Protocols for simultaneous multiple syntheses requiring a minimum of equipment are presented and the use of polyethylene glycol-bound peptides in biochemical binding and functional assay systems is demonstrated.
Sigma-Aldrich
O-Methyl-O′-succinylpolyethylene glycol 2′000
Pricing and availability is not currently available.
Sigma-Aldrich
O-Methyl-O′-succinylpolyethylene glycol 5′000
Pricing and availability is not currently available.