Merck
  • PBRM1 acts as a p53 lysine-acetylation reader to suppress renal tumor growth.
PBRM1 acts as a p53 lysine-acetylation reader to suppress renal tumor growth.
Nature communications (2019-12-22)
Weijia Cai, Liya Su, Lili Liao, Zongzhi Z Liu, Lauren Langbein, Essel Dulaimi, Joseph R Testa, Robert G Uzzo, Zhijiu Zhong, Wei Jiang, Qin Yan, Qing Zhang, Haifeng Yang
ABSTRACT
p53 acetylation is indispensable for its transcriptional activity and tumor suppressive function. However, the identity of reader protein(s) for p53 acetylation remains elusive. PBRM1, the second most highly mutated tumor suppressor gene in kidney cancer, encodes PBRM1. Here, we identify PBRM1 as a reader for p53 acetylation on lysine 382 (K382Ac) through its bromodomain 4 (BD4). Notably, mutations on key residues of BD4 disrupt recognition of p53 K382Ac. The mutation in BD4 also reduces p53 binding to promoters of target genes such as CDKN1A (p21). Consequently, the PBRM1 BD4 mutant fails to fully support p53 transcriptional activity and is defective as a tumor suppressor. We also find that expressions of PBRM1 and p21 correlate with each other in human kidney cancer samples. Our findings uncover a tumor suppressive mechanism of PBRM1 in kidney cancer and provide a mechanistic insight into the crosstalk between p53 and SWI/SNF complexes.
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MDM2 (Ab-5) Mouse mAb (4B2C1.11), liquid, clone 4B2C1.11, Calbiochem®
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse