Merck
  • Organic charge transfer complexes on graphene with ultrahigh near infrared photogain.
Organic charge transfer complexes on graphene with ultrahigh near infrared photogain.
Nanotechnology (2019-02-12)
Muhammad Ahsan Iqbal, Menghua Cui, Adeel Liaqat, Rakba Faiz, Mongur Hossain, Xinsheng Wang, Sabir Hussain, Chunhe Dang, Haining Liu, Wen Wen, Junxia Wu, Liming Xie
ABSTRACT
Photodetectors have widespread applications in fields including telecommunications, thermal imaging and bio-medical imaging. The photogating effect, arising from charge trapping at defects and/or interfaces, can have extremely high photoelectric gain which can be a benefit to high-sensitivity room temperature photodetection. Here, we introduce thin layered organic charge transfer complexes (CPXs) integrated on graphene transistors for the development of hybrid phototransistors with ultra-high photoresponsivity of ∼106 A W-1 in the near infrared (NIR) region at room temperature. Our study has demonstrated a graphene-organic CPX with a broadband photoresponse ranging from the visible to the NIR region. The high photoelectric gain was from the photogating effect at the graphene/CPX interface. In addition, the photoresponse properties of the graphene-organic CPX can be regulated by electrical gating of graphene.
Sigma-Aldrich
3,3′′′-Dihexyl-2,2′:5′,2′′:5′′,2′′′-quaterthiophene, 95%
Sigma-Aldrich
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 97%
Sigma-Aldrich
2,2′:5′,2′′:5′′,2′′′-Quaterthiophene, 96%
Sigma-Aldrich
Poly(3-hexylthiophene-2,5-diyl), regiorandom